Assay Details
Target Gene Details
Entrez Gene ID: | 217207 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 8 |
Gene Aliases: |
Ddx8, mDEAH6, mKIAA4096 |
Location: |
Chr.11:101732956-101767358 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dhx8 | NM_144831.2 | 23 | 4197 | NP_659080.2 |
AK142226.1 | 23 | 4131 | BAE24985.1 | |
AK170616.1 | 23 | 4130 | ||
AK220485.1 | 23 | 4174 | BAD90286.1 | |
BC022656.1 | 6 | 1402 | AAH22656.1 | |
BC132445.1 | 23 | 4181 |
More Information
Set Membership: |
Intragenic Exonic |