Assay Details
Target Gene Details
Entrez Gene ID: | 20303 |
Gene Name: | chemokine (C-C motif) ligand 4 |
Gene Aliases: |
AT744.1, Act-2, MIP-1B, Mip1b, Scya4 |
Location: |
Chr.11:83648624-83664683 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl4 | NM_013652.2 | 3 | 458 | NP_038680.1 |
AF128218.1 | 3 | 431 | AAF22559.1 | |
AF128219.1 | 3 | 431 | AAF22560.1 | |
AK156948.1 | 3 | 451 | BAE33909.1 | |
AK172472.1 | 3 | 451 | BAE43025.1 | |
BC119257.1 | 3 | 405 | ||
BC119259.1 | 3 | 405 | ||
BY157376.1 | 1 | 116 | ||
M23503.1 | 3 | 456 | AAA40148.1 | |
M35590.1 | 3 | 441 | AAA39708.1 |
More Information
Set Membership: |
Intragenic Exonic |