Assay Details
Target Gene Details
Entrez Gene ID: | 26897 |
Gene Name: | acyl-CoA thioesterase 1 |
Gene Aliases: |
ACH2, CTE-1, CTE-I, Cte1, D12Ucla1 |
Location: |
Chr.12:84009502-84017670 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acot1 | NM_012006.2 | 3 | 1257 | NP_036136.1 |
BC003911.1 | 3 | 1254 | AAH03911.1 | |
BC028261.1 | 3 | 1267 | AAH28261.1 | |
Y14004.1 | 3 | 1257 | CAA74327.1 |
More Information
Set Membership: |
Intragenic Exonic |