Assay Details
Target Gene Details
Entrez Gene ID: | 20682 |
Gene Name: | SRY (sex determining region Y)-box 9 |
Gene Aliases: |
2010306G03Rik, AV220920, mKIAA4243 |
Location: |
Chr.11:112782210-112787757 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sox9 | NM_011448.4 | 2 | 1012 | NP_035578.3 |
AF421878.1 | 2 | 637 | AAL16093.1 | |
AK030187.1 | 2 | 997 | BAC26830.1 | |
AK031516.1 | 2 | 996 | ||
AK049986.1 | 2 | 998 | BAC34018.1 | |
AK220197.1 | 2 | 982 | BAD90382.1 | |
BC023796.1 | 2 | 999 | AAH23796.1 | |
BC023808.1 | 2 | 1000 | AAH23808.1 | |
BC023953.1 | 2 | 1009 | AAH23953.1 | |
BC024958.1 | 2 | 989 | AAH24958.1 | |
BC034264.1 | 2 | 940 | AAH34264.1 |