Assay Details
Target Gene Details
Entrez Gene ID: | 17134 |
Gene Name: | v-maf musculoaponeurotic fibrosarcoma oncogene family, protein G (avian) |
Gene Aliases: |
AA545192, C630022N07Rik |
Location: |
Chr.11:120628348-120633547 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mafg | NM_010756.3 | 3 | 1175 | NP_034886.1 |
AK218693.1 | 1 | 162 | ||
BC002092.1 | 3 | 1113 | AAH02092.1 | |
BC052633.1 | 3 | 1175 | AAH52633.1 |
More Information
Set Membership: |
Intragenic Exonic |