Assay Details
Target Gene Details
Entrez Gene ID: | 16199 |
Gene Name: | interleukin 9 receptor |
Gene Aliases: |
- |
Location: |
Chr.11:32187544-32205340 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il9r | NM_001134458.1 | 9 | 2414 | NP_001127930.1 |
NM_008374.2 | 9 | 2411 | NP_032400.1 | |
M84746.1 | 9 | 2354 | AAA37871.1 |
More Information
Set Membership: |
Intragenic Exonic |