Assay Details
Target Gene Details
Entrez Gene ID: | 72047 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 42 |
Gene Aliases: |
1810047H21Rik, AW319508, AW556242, B430002H05Rik, RHELP, RNAHP, SF3b125 |
Location: |
Chr.11:106216463-106249140 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx42 | NM_028074.4 | 14 | 1735 | NP_082350.3 |
AK007805.1 | BAB25270.3 | |||
AK049311.1 | 14 | 1737 | BAC33675.1 | |
AK134182.1 | 13 | 1541 | ||
AK169816.1 | 14 | 1681 | BAE41388.1 | |
AK171730.1 | 14 | 1604 | BAE42635.1 | |
BC043036.1 | 13 | 1492 |