Assay Details
Target Gene Details
Entrez Gene ID: | 11932 |
Gene Name: | ATPase, Na+/K+ transporting, beta 2 polypeptide |
Gene Aliases: |
Amog, Atpb-2 |
Location: |
Chr.11:69599750-69605960 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp1b2 | NM_013415.5 | 7 | 2129 | NP_038201.1 |
AK141786.1 | 7 | 2130 | BAE24833.1 | |
AK158321.1 | 7 | 2132 | ||
BC042467.1 | 7 | 1998 | AAH42467.1 | |
BC058763.1 | 7 | 2087 | AAH58763.1 |
More Information
Set Membership: |
Intragenic Exonic |