Assay Details
Target Gene Details
Entrez Gene ID: | 71275 |
Gene Name: | NADP+ dependent oxidoreductase domain containing 1 |
Gene Aliases: |
4933437F05Rik |
Location: |
Chr.12:87221123-87238774 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Noxred1 | NM_027744.1 | 8 | 1144 | NP_082020.1 |
AK017096.1 | 8 | 1275 | BAB30594.1 | |
BC125331.1 | 8 | 1144 | ||
BC125333.1 | 8 | 1144 |
More Information
Set Membership: |
Intragenic Exonic |