Assay Details
Target Gene Details
Entrez Gene ID: | 76113 |
Gene Name: | lactoperoxidase |
Gene Aliases: |
5830499B15Rik |
Location: |
Chr.11:87806428-87826135 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lpo | NM_080420.2 | 10 | 1805 | NP_536345.2 |
AK018070.1 | 2 | 289 | ||
AK162992.1 | 2 | 253 | BAE37148.1 | |
BC016212.1 | 10 | 1812 | AAH16212.1 |
More Information
Set Membership: |
Intragenic Exonic |