Assay Details
Target Gene Details
Entrez Gene ID: | 276852 |
Gene Name: | DNA segment, Chr 11, Wayne State University 47, expressed |
Gene Aliases: |
- |
Location: |
Chr.11:113684400-113727444 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
D11Wsu47e | NM_177777.5 | 1 | 55 | NP_808445.2 |
AK017508.1 | BAB30780.1 | |||
AK139389.1 | 1 | 28 | BAE23989.1 | |
AK171307.1 | 1 | 63 | BAE42383.1 | |
BC048942.1 | 1 | 58 | AAH48942.1 |