Assay Details
Target Gene Details
Entrez Gene ID: | 103711 |
Gene Name: | pyridoxine 5'-phosphate oxidase |
Gene Aliases: |
AI415282 |
Location: |
Chr.11:96937816-96944019 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pnpo | NM_134021.2 | 7 | 985 | NP_598782.1 |
AK154004.1 | 7 | 925 | BAE32309.1 | |
AK154443.1 | 7 | 746 | BAE32590.1 | |
BC010785.1 | 7 | 924 | AAH10785.1 | |
BC026564.1 | 7 | 952 | AAH26564.1 |
More Information
Set Membership: |
Intragenic Exonic |