Assay Details
Target Gene Details
Entrez Gene ID: | 12418 |
Gene Name: | chromobox 4 |
Gene Aliases: |
MPc2, PC2 |
Location: |
Chr.11:119077571-119086237 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cbx4 | NM_007625.2 | 5 | 2835 | NP_031651.2 |
AK155152.1 | ||||
AK155512.1 | BAE33302.1 | |||
AK164760.1 | 1 | 599 |
More Information
Set Membership: |
Intragenic Exonic |