Assay Details
Target Gene Details
Entrez Gene ID: | 12418 |
Gene Name: | chromobox 4 |
Gene Aliases: |
MPc2, PC2 |
Location: |
Chr.11:119077571-119086237 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cbx4 | NM_007625.2 | 1 | 243 | NP_031651.2 |
BC045162.1 | 1 | 227 | ||
BC117801.1 | 1 | 117 | ||
U63387.1 | 1 | 155 | AAB96874.1 |
More Information
Set Membership: |
Intragenic Exonic |