Assay Details
Target Gene Details
Entrez Gene ID: | 70419 |
Gene Name: | RIKEN cDNA 2810408A11 gene |
Gene Aliases: |
1700019L01Rik, 4930470J18Rik |
Location: |
Chr.11:69897358-69901721 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
2810408A11Rik | NM_027419.3 | 1 | 344 | NP_081695.2 |
AK013042.1 | 1 | 346 | BAB28617.1 | |
AK015537.1 | 1 | 273 | ||
AK051029.1 | 1 | 316 | ||
AK160144.1 | 1 | 253 | ||
AK171674.1 | 1 | 275 | BAE42603.1 | |
BC051546.1 | 1 | 305 | ||
BC069874.1 | 1 | 344 | AAH69874.1 |
More Information
Set Membership: |
Intragenic Exonic |