Assay Details
Target Gene Details
Entrez Gene ID: | 276891 |
Gene Name: | T cell immunoglobulin and mucin domain containing 4 |
Gene Aliases: |
B430010N18Rik, TIM-4, Tim4 |
Location: |
Chr.11:46810786-46844333 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Timd4 | NM_178759.4 | 2 | 356 | NP_848874.3 |
AK046560.1 | 2 | 355 | BAC32787.1 | |
AK079787.1 | 2 | 355 | BAC37750.1 | |
AK080042.1 | 2 | 355 | BAC37813.1 | |
AK080212.1 | 2 | 357 | BAC37848.1 | |
AY376716.1 | 2 | 355 | AAR23483.1 | |
AY376717.1 | 2 | 355 | AAR23484.1 |
More Information
Set Membership: |
Intragenic Exonic |