Assay Details
Target Gene Details
Entrez Gene ID: | 80981 |
Gene Name: | ADP-ribosylation factor-like 4D |
Gene Aliases: |
1110036H21Rik, ARL6, AW456149, Arf4l, Arfl4, Arl5 |
Location: |
Chr.11:101665541-101667832 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arl4d | NM_025404.3 | 2 | 527 | NP_079680.1 |
AF312686.1 | 1 | 484 | AAG53667.1 | |
AK004126.1 | 2 | 508 | BAB23183.1 | |
AK013320.1 | 2 | 505 | BAB28789.1 | |
BC016113.1 | 2 | 493 | AAH16113.1 | |
BQ830332.1 | 2 | 527 |
More Information
Set Membership: |
Intragenic Exonic |