Assay Details
Target Gene Details
Entrez Gene ID: | 22344 |
Gene Name: | vascular endothelial zinc finger 1 |
Gene Aliases: |
AI848691, AW046909, db1 |
Location: |
Chr.11:88068256-88084729 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vezf1 | NM_016686.4 | 6 | 1814 | NP_057895.2 |
AF104410.3 | 6 | 1814 | AAD02883.1 | |
AK160290.1 |
More Information
Set Membership: |
Intragenic Exonic |