Assay Details
Target Gene Details
Entrez Gene ID: | 327978 |
Gene Name: | schlafen 5 |
Gene Aliases: |
AI956840 |
Location: |
Chr.11:82951349-82964850 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slfn5 | NM_183201.4 | 6 | 3360 | NP_899024.3 |
AK036486.1 | 6 | 3361 | BAC29447.1 | |
BC117909.1 | 5 | 3240 | ||
BC117910.1 | 5 | 3240 |