Assay Details
Target Gene Details
Entrez Gene ID: | 237625 |
Gene Name: | phospholipase A2, group III |
Gene Aliases: |
- |
Location: |
Chr.11:3487596-3499346 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g3 | NM_172791.2 | 1 | 308 | NP_766379.2 |
AK033572.1 | 1 | 252 | BAC28367.1 | |
AK081075.1 | 1 | 236 | BAC38128.1 | |
BC079556.1 | 1 | 308 | AAH79556.1 |