Assay Details
Target Gene Details
Entrez Gene ID: | 19039 |
Gene Name: | lectin, galactoside-binding, soluble, 3 binding protein |
Gene Aliases: |
90K, CyCAP, MAC-2BP, Ppicap, Tango10b |
Location: |
Chr.11:118392752-118401931 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lgals3bp | NM_011150.2 | 6 | 1032 | NP_035280.1 |
AK079915.1 | 6 | 1061 | BAC37782.1 | |
AK137836.1 | 6 | 1009 | BAE23505.1 | |
AK150021.1 | 6 | 1061 | BAE29246.1 | |
AK152182.1 | 6 | 974 | BAE31013.1 | |
AK153313.1 | 6 | 1048 | BAE31894.1 | |
AK153396.1 | 6 | 1050 | BAE31958.1 | |
AK155031.1 | 6 | 974 | BAE33003.1 | |
BC090658.1 | 6 | 1028 | ||
L16894.1 | 6 | 1039 | AAA37499.1 | |
X67809.1 | 6 | 1032 | CAA48010.1 |