Assay Details
Target Gene Details
Entrez Gene ID: | 83560 |
Gene Name: | testis expressed gene 14 |
Gene Aliases: |
C85585 |
Location: |
Chr.11:87405050-87555823 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tex14 | NM_001199293.1 | 14 | 2260 | NP_001186222.1 |
NM_031386.2 | 14 | 2241 | NP_113563.2 | |
AK139808.1 | BAE24144.1 |