Assay Details
Target Gene Details
Entrez Gene ID: | 70373 |
Gene Name: | G patch domain containing 2 like |
Gene Aliases: |
1700020O03Rik, 4732487G21Rik, A630090I05Rik, Kiaa1152 |
Location: |
Chr.12:86241838-86291793 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpatch2l | NM_027405.2 | 10 | 3056 | NP_081681.2 |
AK031712.1 | 9 | 2965 | BAC27527.1 | |
AK129299.1 | 10 | 3033 | BAC98109.1 | |
AK147916.1 | 10 | 3045 | BAE28226.1 | |
BC048169.1 | 10 | 3065 | AAH48169.1 | |
BC058256.1 | 10 | 3056 | AAH58256.1 |
More Information
Set Membership: |
Intragenic Exonic |