Assay Details
Target Gene Details
Entrez Gene ID: | 75870 |
Gene Name: | testicular cell adhesion molecule 1 |
Gene Aliases: |
4930570F09Rik, AI429105 |
Location: |
Chr.11:106273079-106288158 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tcam1 | NM_029467.3 | 6 | 1280 | NP_083743.2 |
AK016263.1 | 6 | 1150 | BAB30167.1 | |
AK076581.1 | 6 | 1201 |