Assay Details
Target Gene Details
Entrez Gene ID: | 268448 |
Gene Name: | PHD finger protein 12 |
Gene Aliases: |
2410142K10Rik, F630045O13, PF1, mKIAA1523 |
Location: |
Chr.11:77982754-78030539 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phf12 | NM_174852.3 | 15 | 3572 | NP_777277.2 |
AK129383.1 | 12 | 2867 | BAC98193.2 | |
AK155354.1 | 15 | 3613 | BAE33212.1 | |
BC043080.1 | 15 | 3175 | AAH43080.1 |
More Information
Set Membership: |
Intragenic Exonic |