Assay Details
Target Gene Details
Entrez Gene ID: | 76719 |
Gene Name: | KAT8 regulatory NSL complex subunit 1 |
Gene Aliases: |
1700081L11Rik, 9430041J06Rik, mKIAA1267 |
Location: |
Chr.11:104333229-104469507 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kansl1 | NM_001081045.1 | 2 | 797 | NP_001074514.1 |
AK037800.1 | 3 | 363 | BAE20523.1 | |
AK122484.1 | 2 | 803 | BAC65766.1 | |
AK153679.1 | 2 | 778 | BAE32141.1 | |
AK161514.1 | 2 | 312 | BAE36437.1 | |
BC043121.1 | 2 | 346 | ||
BC053389.1 | 2 | 347 | ||
BC054752.1 | 3 | 620 | ||
BC079594.1 | 2 | 345 | AAH79594.1 |