Assay Details
Target Gene Details
Entrez Gene ID: | 76719 |
Gene Name: | KAT8 regulatory NSL complex subunit 1 |
Gene Aliases: |
1700081L11Rik, 9430041J06Rik, mKIAA1267 |
Location: |
Chr.11:104333229-104469507 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kansl1 | NM_001081045.1 | 14 | 4191 | NP_001074514.1 |
AK122484.1 | 14 | 4197 | BAC65766.1 | |
AK161032.1 | 3 | 1963 | ||
BC025052.1 | 4 | 1072 | AAH25052.1 | |
BC043121.1 | 14 | 3740 | ||
BC054752.1 | 15 | 4014 | ||
BC079594.1 | 14 | 3739 | AAH79594.1 |