Assay Details
Target Gene Details
Entrez Gene ID: | 74190 |
Gene Name: | exocyst complex component 3-like 4 |
Gene Aliases: |
1200009I06Rik, 1600013K19Rik |
Location: |
Chr.12:111417430-111431680 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Exoc3l4 | NM_001289487.1 | 5 | 1362 | NP_001276416.1 |
NM_001289488.1 | 5 | 1113 | NP_001276417.1 | |
NM_001289489.1 | 4 | 1008 | NP_001276418.1 | |
NM_028807.4 | 4 | 1257 | NP_083083.3 | |
AK004674.1 | 5 | 1114 | ||
AK005443.1 | 3 | 1004 | BAB24037.1 | |
AK154875.1 | 5 | 1776 | BAE32895.1 | |
BC042799.1 | AAH42799.1 | |||
BC075664.1 | 4 | 1098 | AAH75664.1 |
More Information
Set Membership: |
Intragenic Exonic |