Assay Details
Target Gene Details
Entrez Gene ID: | 74190 |
Gene Name: | exocyst complex component 3-like 4 |
Gene Aliases: |
1200009I06Rik, 1600013K19Rik |
Location: |
Chr.12:111417430-111431680 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Exoc3l4 | NM_001289487.1 | 1 | 110 | NP_001276416.1 |
NM_001289488.1 | NP_001276417.1 | |||
NM_001289489.1 | NP_001276418.1 | |||
NM_028807.4 | 1 | 110 | NP_083083.3 | |
AK004674.1 | ||||
AK154875.1 | 1 | 524 | BAE32895.1 | |
BY205345.1 | 1 | 113 | ||
CF915432.1 |
More Information
Set Membership: |
Intragenic Exonic |