Assay Details
Target Gene Details
Entrez Gene ID: | 268470 |
Gene Name: | ubiquitin-conjugating enzyme E2Z (putative) |
Gene Aliases: |
AW049199, C030047H17Rik, D11Moh35 |
Location: |
Chr.11:96047431-96065364 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ube2z | NM_172300.3 | 1 | 348 | NP_758504.3 |
AK077793.1 | 1 | 346 | BAC37014.1 | |
AK149770.1 | 1 | 348 | BAE29074.1 | |
AK152529.1 | 1 | 62 | ||
AK153328.1 | 1 | 79 | BAE31907.1 | |
AK168942.1 | 1 | 41 | BAE40750.1 | |
BC054412.1 | 1 | 51 | AAH54412.1 | |
CA317477.1 | 1 | 372 |