Assay Details
Target Gene Details
Entrez Gene ID: | 215332 |
Gene Name: | solute carrier family 36 (proton/amino acid symporter), member 3 |
Gene Aliases: |
PAT3, TRAMD2, tramdorin2 |
Location: |
Chr.11:55124815-55151745 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc36a3 | NM_172258.3 | 11 | 1589 | NP_758462.2 |
AF453745.1 | 11 | 1589 | AAN76273.1 | |
AK132910.1 | 12 | 1675 | ||
AY211261.1 | 11 | 1571 | AAO37089.1 |
More Information
Set Membership: |
Intragenic Exonic |