Assay Details
Target Gene Details
Entrez Gene ID: | 18102 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 1 |
Gene Aliases: |
AL024257, NDPK-A, NM23-M1, NM23A |
Location: |
Chr.11:93958925-93968521 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nme1 | NM_008704.2 | 5 | 1144 | NP_032730.1 |
U85511.1 | 5 | 931 | AAB42080.1 |
More Information
Set Membership: |
Intragenic Exonic |