Assay Details
Target Gene Details
Entrez Gene ID: | 238023 |
Gene Name: | hexosaminidase (glycosyl hydrolase family 20, catalytic domain) containing |
Gene Aliases: |
BC069960 |
Location: |
Chr.11:121204433-121222656 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hexdc | NM_001001333.2 | NP_001001333.2 | ||
NM_001146073.1 | 9 | 1075 | NP_001139545.1 | |
AK154237.1 | BAE32455.1 | |||
BC069960.1 | 9 | 1027 | AAH69960.1 | |
FM204885.1 | ||||
FM204886.1 | 9 | 1026 |