Assay Details
Target Gene Details
Entrez Gene ID: | 238023 |
Gene Name: | hexosaminidase (glycosyl hydrolase family 20, catalytic domain) containing |
Gene Aliases: |
BC069960 |
Location: |
Chr.11:121204433-121222656 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hexdc | NM_001001333.2 | 1 | 271 | NP_001001333.2 |
AK154237.1 | 1 | 245 | BAE32455.1 | |
BY206924.1 | ||||
BY242213.1 | 1 | 272 | ||
BY263070.1 | 1 | 271 | ||
BY296469.1 |