Assay Details
Target Gene Details
Entrez Gene ID: | 13861 |
Gene Name: | eosinophil peroxidase |
Gene Aliases: |
EPO |
Location: |
Chr.11:87863997-87875536 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epx | NM_007946.2 | 7 | 982 | NP_031972.2 |
AK132416.1 | 2 | 107 | BAE21157.1 | |
D78353.1 | 7 | 988 | BAA11370.1 | |
L77979.1 | 7 | 966 | AAB40403.1 |
More Information
Set Membership: |
Intragenic Exonic |