Assay Details
Target Gene Details
Entrez Gene ID: | 216846 |
Gene Name: | centrobin, centrosomal BRCA2 interacting protein |
Gene Aliases: |
9830165K03Rik, Lip8 |
Location: |
Chr.11:69299491-69323893 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cntrob | NM_172560.3 | 13 | 2762 | NP_766148.2 |
AK036710.1 | 12 | 2582 | BAC29545.1 | |
AK147774.1 | 13 | 2652 | ||
BC058512.1 | 13 | 1595 | AAH58512.1 |