Assay Details
Target Gene Details
Entrez Gene ID: | 20185 |
Gene Name: | nuclear receptor co-repressor 1 |
Gene Aliases: |
5730405M06Rik, A230020K14Rik, N-CoR, RIP13, Rxrip13, mKIAA1047 |
Location: |
Chr.11:62315643-62457332 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ncor1 |
Target Gene Details
Entrez Gene ID: | 72795 |
Gene Name: | tetratricopeptide repeat domain 19 |
Gene Aliases: |
2010204O13Rik, 2810460C24Rik, AI505442 |
Location: |
Chr.11:62281427-62316441 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc19 | NM_028360.2 | 10 | 3019 | NP_082636.3 |
NM_029704.2 | 9 | 2980 | NP_083980.2 | |
AA466515.1 | 1 | 418 | ||
AK034075.1 | 10 | 3009 | BAC28572.1 | |
BB078627.2 | 1 | 261 |