Assay Details
Target Gene Details
Entrez Gene ID: | 15171 |
Gene Name: | hypocretin |
Gene Aliases: |
PPOX |
Location: |
Chr.11:100761693-100762931 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hcrt | NM_010410.2 | 2 | 434 | NP_034540.1 |
AF019566.1 | 2 | 435 | AAC02934.1 | |
AF041242.1 | 2 | 434 | AAC40040.1 | |
BC116943.1 | 2 | 427 | ||
BC116969.1 | 2 | 427 |