Assay Details
Target Gene Details
Entrez Gene ID: | 20892 |
Gene Name: | stimulated by retinoic acid 13 |
Gene Aliases: |
- |
Location: |
Chr.11:120710941-120713903 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stra13 | NM_016665.2 | 5 | 333 | NP_057874.2 |
AK076181.1 | 4 | 392 | BAC36237.1 | |
AK080500.1 | 5 | 305 | BAC37933.1 | |
AK170126.1 | 5 | 305 | BAE41581.1 | |
CA320983.1 | 4 | 272 | ||
CJ236047.1 | 4 | 245 | ||
U95003.1 | 6 | 397 | AAB53635.1 | |
U95004.1 | 5 | 298 | AAB53636.1 | |
U95005.1 | 6 | 571 | AAB53637.1 |