Assay Details
Target Gene Details
Entrez Gene ID: | 20851 |
Gene Name: | signal transducer and activator of transcription 5B |
Gene Aliases: |
- |
Location: |
Chr.11:100780731-100859004 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stat5b | NM_001113563.1 | 19 | 3681 | NP_001107035.1 |
NM_011489.3 | 19 | 4013 | NP_035619.3 | |
AK004071.1 | 1 | 1207 | BAB23152.1 | |
AK045531.1 | 19 | 3824 | ||
AK150098.1 | 19 | 3650 | BAE29305.1 | |
AK154664.1 | 19 | 3647 | BAE32752.1 | |
AK191910.1 | 1 | 167 | ||
BC024319.1 | 19 | 3637 | AAH24319.1 | |
U21110.2 | 19 | 3967 | AAC52282.1 |
More Information
Set Membership: |
Intragenic Exonic |