Assay Details
Target Gene Details
Entrez Gene ID: | 20415 |
Gene Name: | sex hormone binding globulin |
Gene Aliases: |
ABP |
Location: |
Chr.11:69610281-69618025 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Shbg | NM_011367.2 | 8 | 1152 | NP_035497.1 |
AK010959.1 | 8 | 1151 | ||
BC120785.1 | 8 | 1137 | ||
BC120787.1 | 8 | 1137 | ||
U85644.1 | 8 | 1115 | AAB42088.1 |