Assay Details
Target Gene Details
Entrez Gene ID: | 54351 |
Gene Name: | elongator acetyltransferase complex subunit 5 |
Gene Aliases: |
Derp6, Rai12 |
Location: |
Chr.11:69968224-69981294 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Elp5 | NM_001253700.1 | 7 | 854 | NP_001240629.1 |
NM_018740.2 | 9 | 1336 | NP_061210.2 | |
AF004107.1 | 8 | 1255 | AAC16252.1 | |
AK011130.1 | 7 | 1148 | ||
AK013662.1 | 7 | 937 | BAB28944.1 | |
AK152582.1 | 9 | 1340 | BAE31332.1 | |
AK207822.1 | 1 | 185 | ||
AK215451.1 | 2 | 252 | ||
BC003752.1 | 8 | 1187 | AAH03752.1 | |
CD741073.1 | 1 | 116 | ||
CD776986.1 | 1 | 116 |