Assay Details
Target Gene Details
Entrez Gene ID: | 52626 |
Gene Name: | CDKN2A interacting protein N-terminal like |
Gene Aliases: |
A430101B06Rik, AA409260, D11Ertd497e, MNCb-1520 |
Location: |
Chr.11:51967631-51977336 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdkn2aipnl | NM_029976.3 | 3 | 1380 | NP_084252.1 |
AB041653.1 | 3 | 1348 | BAB93546.1 | |
AK077515.1 | 3 | 1205 | ||
AK150573.1 | 3 | 1336 | BAE29668.1 | |
AK171740.1 | 3 | 1315 | BAE42643.1 | |
AK183587.1 | ||||
AK194495.1 | ||||
AK199525.1 |
More Information
Set Membership: |
Intragenic Exonic |