Assay Details
Target Gene Details
Entrez Gene ID: | 66983 |
Gene Name: | zinc finger protein 830 |
Gene Aliases: |
2410003C20Rik, AV301118, AW048207, Ccdc16, Omcg1, Znf830 |
Location: |
Chr.11:82764345-82767622 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp830 | NM_025884.4 | 1 | 1090 | NP_080160.2 |
AK008475.1 | 1 | 1092 | ||
AK010353.1 | 1 | 1081 | BAB26873.1 | |
AK017640.1 | 1 | 1070 | BAB30851.1 | |
AK050043.1 | 1 | 1085 | BAC34045.1 | |
AK132497.1 | 1 | 1091 | BAE21204.1 | |
AK148058.1 | 1 | 1091 | BAE28318.1 | |
AK163064.1 | 1 | 152 | BAE37178.1 | |
AK167656.1 | 1 | 1081 | BAE39707.1 | |
AK168417.1 | 1 | 1070 | BAE40331.1 | |
BC024340.1 | 1 | 1076 | AAH24340.1 |