Assay Details
Target Gene Details
Entrez Gene ID: | 320398 |
Gene Name: | leucine-rich repeats and immunoglobulin-like domains 3 |
Gene Aliases: |
9030421L11Rik, 9130004I02Rik, 9430095K15Rik, mKIAA3016 |
Location: |
Chr.10:125966219-126015359 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrig3 | NM_177152.5 | 14 | 2109 | NP_796126.4 |
AK018522.1 | 8 | 922 | ||
AK129481.1 | 10 | 1058 | BAC98291.1 | |
AY505341.1 | 14 | 2052 | AAR98630.1 | |
BC065142.1 | 14 | 2075 | AAH65142.1 |
More Information
Set Membership: |
Intragenic Exonic |