Assay Details
Target Gene Details
Entrez Gene ID: | 68094 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 2 |
Gene Aliases: |
5930405J04Rik |
Location: |
Chr.10:128459236-128490583 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smarcc2 | NM_001114096.1 | 21 | 2229 | NP_001107568.1 |
NM_001114097.1 | 20 | 2136 | NP_001107569.1 | |
NM_198160.2 | 20 | 2136 | NP_937803.1 | |
AK089461.1 | 20 | 2072 | ||
AK146971.1 | 21 | 2158 | BAE27576.1 | |
AK170530.1 | 20 | 2071 | ||
BC058720.1 | 20 | 2078 | AAH58720.1 | |
BC062102.1 | 9 | 935 | AAH62102.1 |
More Information
Set Membership: |
Intragenic Exonic |