Assay Details
Target Gene Details
Entrez Gene ID: | 432450 |
Gene Name: | Na+/K+ transporting ATPase interacting 2 |
Gene Aliases: |
6330571D19Rik, AW455467, Tcba1 |
Location: |
Chr.10:31689310-32893997 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nkain2 | NM_001013411.2 | 4 | 619 | NP_001013429.2 |
NM_001025286.2 | 4 | 619 | NP_001020457.2 | |
AY940478.1 | 4 | 415 | AAX33293.1 | |
BC141063.1 | 4 | 489 | ||
DQ065606.1 | 4 | 552 | AAY68415.1 | |
DQ065607.1 | 4 | 552 | AAY68416.1 | |
EF058049.2 | 4 | 552 | ||
EF058050.1 | 3 | 400 | ||
EF058051.1 | 4 | 400 |