Assay Details
Target Gene Details
Entrez Gene ID: | 432450 |
Gene Name: | Na+/K+ transporting ATPase interacting 2 |
Gene Aliases: |
6330571D19Rik, AW455467, Tcba1 |
Location: |
Chr.10:31689310-32893997 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nkain2 | NM_001013411.2 | 1 | 139 | NP_001013429.2 |
NM_001025286.2 | 1 | 139 | NP_001020457.2 | |
AK029229.1 | 1 | 686 | BAC26355.1 | |
AK142002.1 | 1 | 719 | ||
BC141063.1 | ||||
DQ065606.1 | 1 | 72 | AAY68415.1 | |
DQ065607.1 | 1 | 72 | AAY68416.1 | |
EF058049.2 | 1 | 72 |