Assay Details
Target Gene Details
Entrez Gene ID: | 432450 |
Gene Name: | Na+/K+ transporting ATPase interacting 2 |
Gene Aliases: |
6330571D19Rik, AW455467, Tcba1 |
Location: |
Chr.10:31689310-32893997 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nkain2 | NM_001013411.2 | 7 | 2418 | NP_001013429.2 |
AK018248.1 | 1 | 481 | ||
AY940478.1 | 7 | 2214 | AAX33293.1 | |
EF058049.2 | 7 | 2351 |