Assay Details
Target Gene Details
Entrez Gene ID: | 22295 |
Gene Name: | cadherin 23 (otocadherin) |
Gene Aliases: |
4930542A03Rik, USH1D, ahl, ahl1, bob, bus, mdfw, nmf112, nmf181, nmf252, sals, v |
Location: |
Chr.10:60302414-60696515 on Build GRCm38 |
Assay Gene Location: | Within Exon 62 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh23 | NM_001252635.1 | 61 | 9165 | NP_001239564.1 |
NM_023370.3 | 62 | 9171 | NP_075859.2 | |
NR_045556.1 | 61 | 9165 | ||
AF308939.1 | 60 | 8764 | AAG52817.1 | |
AJ888000.1 | 59 | 8758 | CAI59609.1 | |
AJ888001.1 | 59 | 8758 | CAI59610.1 | |
AJ888002.1 | 59 | 8758 | CAI59611.1 | |
AK039126.1 | 17 | 2506 | ||
AY026062.1 | 60 | 8773 | AAK07670.1 | |
AY563163.1 | 14 | 2386 | AAT72163.1 | |
AY563164.1 | 14 | 2386 | AAT72164.1 |
More Information
Set Membership: |
Intragenic Exonic |